RIE156323261 Rieke Metals, Inc.

2-(4-Isopropylbenzoyl)pyridine

  • MDL number MFCD07699246
  • Cas Number : 206357-76-8
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-isopropylphenyl)(2-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-11(2)12-6-8-13(9-7-12)15(17)14-5-3-4-10-16-14/h3-11H,1-2H3
Inchi Key LADFKBHPBOIEAA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5570-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5570-32
Availability details for
Quantity
  • Availability
  • Add to Cart