RIE156324273 Rieke Metals, Inc.

cyclobutyl 4-(morpholinomethyl)phenyl ketone

  • MDL number MFCD03841430
  • Cas Number : 898770-75-7
  • Molecular Weight : 259.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0102-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0102-00
Availability details for
Quantity
  • Availability
  • Add to Cart