RIE156324280 Rieke Metals, Inc.

ethyl 8-[4-(morpholinomethyl)phenyl]-8-oxooctanoate

  • MDL number MFCD03841437
  • Cas Number : 898770-89-3
  • Molecular Weight : 361.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0109-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0109-00
Availability details for
Quantity
  • Availability
  • Add to Cart