RIE156324282 Rieke Metals, Inc.

2-methyl-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841439
  • Cas Number : 898770-93-9
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-02
Availability details for
Quantity
  • Availability
  • Add to Cart