RIE156324283 Rieke Metals, Inc.

3-methyl-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841440
  • Cas Number : 898770-95-1
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-03
Availability details for
Quantity
  • Availability
  • Add to Cart