RIE156324284 Rieke Metals, Inc.

4-methyl-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841441
  • Cas Number : 898770-97-3
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methylphenyl)[4-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NO/c1-16-5-9-18(10-6-16)20(22)19-11-7-17(8-12-19)15-21-13-3-2-4-14-21/h5-12H,2-4,13-15H2,1H3
Inchi Key MJYNQTOKIIJFCO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0200-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-04
Availability details for
Quantity
  • Availability
  • Add to Cart