RIE156324285 Rieke Metals, Inc.

2-methoxy-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841442
  • Cas Number : 898770-99-5
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methoxyphenyl)[4-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NO2/c1-23-19-8-4-3-7-18(19)20(22)17-11-9-16(10-12-17)15-21-13-5-2-6-14-21/h3-4,7-12H,2,5-6,13-15H2,1H3
Inchi Key BKNOEGQBQUASCF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0200-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-05
Availability details for
Quantity
  • Availability
  • Add to Cart