RIE156324286 Rieke Metals, Inc.

3-methoxy-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841443
  • Cas Number : 898771-01-2
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-06
Availability details for
Quantity
  • Availability
  • Add to Cart