RIE156324292 Rieke Metals, Inc.

3-carboethoxy-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841449
  • Cas Number : 898771-17-0
  • Molecular Weight : 351.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-12
Availability details for
Quantity
  • Availability
  • Add to Cart