RIE156324294 Rieke Metals, Inc.

4'-piperidinomethyl-2-thiomethylbenzophenone

  • MDL number MFCD03841451
  • Cas Number : 898771-23-8
  • Molecular Weight : 325.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-14
Availability details for
Quantity
  • Availability
  • Add to Cart