RIE156324295 Rieke Metals, Inc.

4-piperidinomethyl-4'-thiomethylbenzophenone

  • MDL number MFCD03841452
  • Cas Number : 898771-26-1
  • Molecular Weight : 325.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(methylsulfanyl)phenyl][4-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NOS/c1-23-19-11-9-18(10-12-19)20(22)17-7-5-16(6-8-17)15-21-13-3-2-4-14-21/h5-12H,2-4,13-15H2,1H3
Inchi Key YXLWYKJKSWEDDS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0200-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-15
Availability details for
Quantity
  • Availability
  • Add to Cart