RIE156324296 Rieke Metals, Inc.

3-bromo-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841453
  • Cas Number : 898771-29-4
  • Molecular Weight : 358.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)[4-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H20BrNO/c20-18-6-4-5-17(13-18)19(22)16-9-7-15(8-10-16)14-21-11-2-1-3-12-21/h4-10,13H,1-3,11-12,14H2
Inchi Key UXZNNOKIXLKJBA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0200-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-18
Availability details for
Quantity
  • Availability
  • Add to Cart