RIE156324307 Rieke Metals, Inc.

3,5-dimethyl-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841464
  • Cas Number : 898775-14-9
  • Molecular Weight : 307.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-31
Availability details for
Quantity
  • Availability
  • Add to Cart