RIE156324310 Rieke Metals, Inc.

3-chloro-4-fluoro-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841467
  • Cas Number : 898775-23-0
  • Molecular Weight : 331.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-4-fluorophenyl)[4-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H19ClFNO/c20-17-12-16(8-9-18(17)21)19(23)15-6-4-14(5-7-15)13-22-10-2-1-3-11-22/h4-9,12H,1-3,10-11,13H2
Inchi Key JEDNNHSXQSWDJB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0200-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-34
Availability details for
Quantity
  • Availability
  • Add to Cart