RIE156324312 Rieke Metals, Inc.

2-fluoro-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841469
  • Cas Number : 898775-29-6
  • Molecular Weight : 297.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-fluorophenyl)[4-(1-piperidinylmethyl)phenyl]methanone
Inchi Code 1S/C19H20FNO/c20-18-7-3-2-6-17(18)19(22)16-10-8-15(9-11-16)14-21-12-4-1-5-13-21/h2-3,6-11H,1,4-5,12-14H2
Inchi Key KPSZNZQNDRJODV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0200-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-37
Availability details for
Quantity
  • Availability
  • Add to Cart