RIE156324313 Rieke Metals, Inc.

4'-piperidinomethyl-2-trifluoromethylbenzophenone

  • MDL number MFCD03841470
  • Cas Number : 898775-32-1
  • Molecular Weight : 347.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(1-piperidinylmethyl)phenyl][2-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C20H20F3NO/c21-20(22,23)18-7-3-2-6-17(18)19(25)16-10-8-15(9-11-16)14-24-12-4-1-5-13-24/h2-3,6-11H,1,4-5,12-14H2
Inchi Key MBTOGVFKJWVFTG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0200-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-43
Availability details for
Quantity
  • Availability
  • Add to Cart