RIE156324315 Rieke Metals, Inc.

4-piperidinomethyl-4'-trifluoromethylbenzophenone

  • MDL number MFCD03841472
  • Cas Number : 898775-38-7
  • Molecular Weight : 347.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-45
Availability details for
Quantity
  • Availability
  • Add to Cart