RIE156324318 Rieke Metals, Inc.

3-chloro-5-fluoro-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841475
  • Cas Number : 898775-47-8
  • Molecular Weight : 331.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-51
Availability details for
Quantity
  • Availability
  • Add to Cart