RIE156324327 Rieke Metals, Inc.

3,5-difluoro-4'-piperidinomethyl benzophenone

  • MDL number MFCD03841484
  • Cas Number : 898775-67-2
  • Molecular Weight : 315.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0200-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0200-60
Availability details for
Quantity
  • Availability
  • Add to Cart