RIE156324334 Rieke Metals, Inc.

ethyl 5-oxo-5-[4-(piperidinomethyl)phenyl]valerate

  • MDL number MFCD03841491
  • Cas Number : 898775-81-0
  • Molecular Weight : 317.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-oxo-5-[4-(1-piperidinylmethyl)phenyl]pentanoate
Inchi Code 1S/C19H27NO3/c1-2-23-19(22)8-6-7-18(21)17-11-9-16(10-12-17)15-20-13-4-3-5-14-20/h9-12H,2-8,13-15H2,1H3
Inchi Key BZQJWZJHAIYNAH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0206-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0206-00
Availability details for
Quantity
  • Availability
  • Add to Cart