RIE156324335 Rieke Metals, Inc.

ethyl 6-oxo-6-[4-(piperidinomethyl)phenyl]hexanoate

  • MDL number MFCD03841492
  • Cas Number : 898775-83-2
  • Molecular Weight : 331.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0207-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0207-00
Availability details for
Quantity
  • Availability
  • Add to Cart