RIE156324340 Rieke Metals, Inc.

3-methyl-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841497
  • Cas Number : 898775-93-4
  • Molecular Weight : 279.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0300-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-03
Availability details for
Quantity
  • Availability
  • Add to Cart