RIE156324349 Rieke Metals, Inc.

4-carboethoxy-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841507
  • Cas Number : 898776-17-5
  • Molecular Weight : 337.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-[4-(1-pyrrolidinylmethyl)benzoyl]benzoate
Inchi Code 1S/C21H23NO3/c1-2-25-21(24)19-11-9-18(10-12-19)20(23)17-7-5-16(6-8-17)15-22-13-3-4-14-22/h5-12H,2-4,13-15H2,1H3
Inchi Key BNHWEVFQNBWPSF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0300-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-13
Availability details for
Quantity
  • Availability
  • Add to Cart