RIE156324358 Rieke Metals, Inc.

2,5-dimethyl-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841518
  • Cas Number : 898776-47-1
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0300-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-28
Availability details for
Quantity
  • Availability
  • Add to Cart