RIE156324365 Rieke Metals, Inc.

2-chloro-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841525
  • Cas Number : 898776-61-9
  • Molecular Weight : 299.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl)[4-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C18H18ClNO/c19-17-6-2-1-5-16(17)18(21)15-9-7-14(8-10-15)13-20-11-3-4-12-20/h1-2,5-10H,3-4,11-13H2
Inchi Key PYZCZLHRQATHQA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0300-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-36
Availability details for
Quantity
  • Availability
  • Add to Cart