RIE156324370 Rieke Metals, Inc.

4-bromo-2-fluoro-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841530
  • Cas Number : 898776-71-1
  • Molecular Weight : 362.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-2-fluorophenyl)[4-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17BrFNO/c19-15-7-8-16(17(20)11-15)18(22)14-5-3-13(4-6-14)12-21-9-1-2-10-21/h3-8,11H,1-2,9-10,12H2
Inchi Key JVSKUOXAVUUJDZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0300-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-49
Availability details for
Quantity
  • Availability
  • Add to Cart