RIE156324371 Rieke Metals, Inc.

2-chloro-4-fluoro-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841531
  • Cas Number : 898776-73-3
  • Molecular Weight : 317.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chloro-4-fluorophenyl)[4-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17ClFNO/c19-17-11-15(20)7-8-16(17)18(22)14-5-3-13(4-6-14)12-21-9-1-2-10-21/h3-8,11H,1-2,9-10,12H2
Inchi Key KVIIPMYFVUNDRU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0300-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-50
Availability details for
Quantity
  • Availability
  • Add to Cart