RIE156324380 Rieke Metals, Inc.

3,4-difluoro-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841540
  • Cas Number : 898776-91-5
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-difluorophenyl)[4-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17F2NO/c19-16-8-7-15(11-17(16)20)18(22)14-5-3-13(4-6-14)12-21-9-1-2-10-21/h3-8,11H,1-2,9-10,12H2
Inchi Key AMAQRLIZIIEDHT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0300-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-59
Availability details for
Quantity
  • Availability
  • Add to Cart