RIE156324381 Rieke Metals, Inc.

3,5-difluoro-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841541
  • Cas Number : 898776-93-7
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)[4-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17F2NO/c19-16-9-15(10-17(20)11-16)18(22)14-5-3-13(4-6-14)12-21-7-1-2-8-21/h3-6,9-11H,1-2,7-8,12H2
Inchi Key KRRCOINVEADLGF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0300-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-60
Availability details for
Quantity
  • Availability
  • Add to Cart