RIE156324383 Rieke Metals, Inc.

cyclopropyl 4-(pyrrolidinomethyl)phenyl ketone

  • MDL number MFCD03841543
  • Cas Number : 898776-97-1
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0301-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0301-00
Availability details for
Quantity
  • Availability
  • Add to Cart