RIE156324387 Rieke Metals, Inc.

ethyl 4-oxo-4-[(4-pyrrolidinomthyl)phenyl]butyrate

  • MDL number MFCD03841547
  • Cas Number : 898777-07-6
  • Molecular Weight : 289.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0305-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0305-00
Availability details for
Quantity
  • Availability
  • Add to Cart