RIE156324388 Rieke Metals, Inc.

ethyl 5-oxo-5-[(4-pyrrolidinomthyl)phenyl]valerate

  • MDL number MFCD03841548
  • Cas Number : 898777-10-1
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-oxo-5-[4-(1-pyrrolidinylmethyl)phenyl]pentanoate
Inchi Code 1S/C18H25NO3/c1-2-22-18(21)7-5-6-17(20)16-10-8-15(9-11-16)14-19-12-3-4-13-19/h8-11H,2-7,12-14H2,1H3
Inchi Key BDZZGBACQGNQRO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0306-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0306-00
Availability details for
Quantity
  • Availability
  • Add to Cart