RIE156324389 Rieke Metals, Inc.

ethyl 6-oxo-6-[(4-pyrrolidinomthyl)phenyl]hexanoate

  • MDL number MFCD03841549
  • Cas Number : 898777-13-4
  • Molecular Weight : 317.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0307-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0307-00
Availability details for
Quantity
  • Availability
  • Add to Cart