RIE156324392 Rieke Metals, Inc.

4-(azetidinomethyl) benzophenone

  • MDL number MFCD03841552
  • Cas Number : 898777-22-5
  • Molecular Weight : 251.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-01
Availability details for
Quantity
  • Availability
  • Add to Cart