RIE156324399 Rieke Metals, Inc.

4'-azetidinomethyl-2-cyanobenzophenone

  • MDL number MFCD03841559
  • Cas Number : 898777-43-0
  • Molecular Weight : 276.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-08
Availability details for
Quantity
  • Availability
  • Add to Cart