RIE156324408 Rieke Metals, Inc.

4-azetidinomethyl-4'-bromobenzophenone

  • MDL number MFCD03841568
  • Cas Number : 898756-43-9
  • Molecular Weight : 330.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-19
Availability details for
Quantity
  • Availability
  • Add to Cart