RIE156324411 Rieke Metals, Inc.

4'-azetidinomethyl-3-fluorobenzophenone

  • MDL number MFCD03841571
  • Cas Number : 898756-52-0
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-22
Availability details for
Quantity
  • Availability
  • Add to Cart