RIE156324412 Rieke Metals, Inc.

4-azetidinomethyl-4'-fluorobenzophenone

  • MDL number MFCD03841572
  • Cas Number : 898756-55-3
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-23
Availability details for
Quantity
  • Availability
  • Add to Cart