RIE156324413 Rieke Metals, Inc.

4'-azetidinomethyl-2,3-dimethylbenzophenone

  • MDL number MFCD03841573
  • Cas Number : 898756-58-6
  • Molecular Weight : 279.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-26
Availability details for
Quantity
  • Availability
  • Add to Cart