RIE156324415 Rieke Metals, Inc.

4'-azetidinomethyl-2,5-dimethylbenzophenone

  • MDL number MFCD03841575
  • Cas Number : 898756-64-4
  • Molecular Weight : 279.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-28
Availability details for
Quantity
  • Availability
  • Add to Cart