RIE156324416 Rieke Metals, Inc.

4'-azetidinomethyl-2,6-dimethylbenzophenone

  • MDL number MFCD03841576
  • Cas Number : 898756-67-7
  • Molecular Weight : 279.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0400-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-29
Availability details for
Quantity
  • Availability
  • Add to Cart