RIE156324418 Rieke Metals, Inc.

4'-azetidinomethyl-3,5-dimethylbenzophenone

  • MDL number MFCD03841578
  • Cas Number : 898756-71-3
  • Molecular Weight : 279.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-31
Availability details for
Quantity
  • Availability
  • Add to Cart