RIE156324419 Rieke Metals, Inc.

4'-azetidinomethyl-4-bromo-3-fluorobenzophenone

  • MDL number MFCD03841579
  • Cas Number : 898756-73-5
  • Molecular Weight : 348.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-32
Availability details for
Quantity
  • Availability
  • Add to Cart