RIE156324420 Rieke Metals, Inc.

4'-azetidinomethyl-4-chloro-3-fluorobenzophenone

  • MDL number MFCD03841580
  • Cas Number : 898756-75-7
  • Molecular Weight : 303.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0400-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-33
Availability details for
Quantity
  • Availability
  • Add to Cart