RIE156324422 Rieke Metals, Inc.

4'-azetidinomethyl-2-chlorobenzophenone

  • MDL number MFCD03841582
  • Cas Number : 898756-79-1
  • Molecular Weight : 285.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(1-azetidinylmethyl)phenyl](2-chlorophenyl)methanone
Inchi Code 1S/C17H16ClNO/c18-16-5-2-1-4-15(16)17(20)14-8-6-13(7-9-14)12-19-10-3-11-19/h1-2,4-9H,3,10-12H2
Inchi Key VNJNAOWJSQIXDU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0400-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-36
Availability details for
Quantity
  • Availability
  • Add to Cart