RIE156324424 Rieke Metals, Inc.

4'-azetidinomethyl-2-trifluoromethylbenzophenone

  • MDL number MFCD03841584
  • Cas Number : 898756-82-6
  • Molecular Weight : 319.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-43
Availability details for
Quantity
  • Availability
  • Add to Cart