RIE156324434 Rieke Metals, Inc.

4'-azetidinomethyl-3,4-dichlorobenzophenone

  • MDL number MFCD03841594
  • Cas Number : 898757-00-1
  • Molecular Weight : 320.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0400-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0400-56
Availability details for
Quantity
  • Availability
  • Add to Cart