RIE156324440 Rieke Metals, Inc.

4-(azetidinomethyl)phenyl cyclopropyl ketone

  • MDL number MFCD03841600
  • Cas Number : 898757-10-3
  • Molecular Weight : 215.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0401-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0401-00
Availability details for
Quantity
  • Availability
  • Add to Cart