RIE156324442 Rieke Metals, Inc.

4-(azetidinomethyl)phenyl cyclopentyl ketone

  • MDL number MFCD03841602
  • Cas Number : 898757-14-7
  • Molecular Weight : 243.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0403-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0403-00
Availability details for
Quantity
  • Availability
  • Add to Cart