RIE156324443 Rieke Metals, Inc.

4-(azetidinomethyl)phenyl cyclohexyl ketone

  • MDL number MFCD03841603
  • Cas Number : 898757-16-9
  • Molecular Weight : 257.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(1-azetidinylmethyl)phenyl](cyclohexyl)methanone
Inchi Code 1S/C17H23NO/c19-17(15-5-2-1-3-6-15)16-9-7-14(8-10-16)13-18-11-4-12-18/h7-10,15H,1-6,11-13H2
Inchi Key GZEKLDPISHKODV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0404-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0404-00
Availability details for
Quantity
  • Availability
  • Add to Cart